Learn More
New England Biolabs, Inc. Lambda PFG Ladder – 0.05 ml
SDP
Catalog No. 501526412
Encompass
Lambda PFG Ladder consists of one GelSyringe™ dispenser, sufficient for 50 gel lanes. Successively larger concatemers of lambda DNA (cI857 ind 1 Sam7) are embedded in 1% LMP agarose. This product is designed to be used as size markers for pulsed-field gel electrophoresis (PFG). Size range: 48.5–1,018 kb.
Catalog No. 50-152-6412 Supplier New England Biolabs, Inc. Supplier No. N0341S
Lambda PFG Ladder consists of one GelSyringe™ dispenser, sufficient for 50 gel lanes. Successively larger concatemers of lambda DNA (cI857 ind 1 Sam7) are embedded in 1% LMP agarose. This product is designed to be used as size markers for pulsed-field gel electrophoresis (PFG). Size range: 48.5–1,018 kb.