Learn More
Santa Cruz Biotechnology Lambda Phosphatase, 20000 units
Catalog No. NC0257512
Encompass
Lambda Phosphatase can be used to release phosphate groups from phosphorylated serine, threonine and tyrosine residues in proteins; also active on phosphorylated histidine residues. 1 mL of 10X Reaction buffer and 1 mL of 10X MnCl2 included with each size. Lambda Phosphatase (Lambda PP) is a Mn2+-dependent protein phosphatase with activity towards phosphorylated serine, threonine and tyrosine residues. The enzyme is inhibited by vanadate ions and can be heat inactivated at 65° C for 1 hour in the presence of 50 mM EDTA.
Catalog No. NC0257512 Supplier Santa Cruz Biotechnology Supplier No. SC200312A
Lambda Phosphatase can be used to release phosphate groups from phosphorylated serine, threonine and tyrosine residues in proteins; also active on phosphorylated histidine residues. 1 mL of 10X Reaction buffer and 1 mL of 10X MnCl2 included with each size. Lambda Phosphatase (Lambda PP) is a Mn2+-dependent protein phosphatase with activity towards phosphorylated serine, threonine and tyrosine residues. The enzyme is inhibited by vanadate ions and can be heat inactivated at 65° C for 1 hour in the presence of 50 mM EDTA.