Learn More
New England Biolabs, Inc. Lambda Protein Phosphatase – 20000 units
SDP
Catalog No. 50811856
Encompass
Lambda Protein Phosphatase is a Mn2+-dependent protein phosphatase with activity towards phosphorylated serine, threonine and tyrosine residues.
Catalog No. 50-811-856 Supplier New England Biolabs, Inc. Supplier No. P0753S
Lambda Protein Phosphatase is a Mn2+-dependent protein phosphatase with activity towards phosphorylated serine, threonine and tyrosine residues.