Learn More
Medchemexpress LLC Lamiide | 27856-54-8 | 1 MG

Supplier: Medchemexpress LLC HYN34081MG
Lamiide is an orally active iridoid compound derived from *Bouchea fluminensis* leaves, known for its anti-inflammatory and anti-lipoperoxidative properties. It has been shown to inhibit rat-brain phospholipid peroxidation with an IC50 of 0.92 mM *in vitro*. *In vivo* studies indicate that Lamiide dose-dependently reduces carrageenin-induced edema in female Wistar rats, with an ED50 of 62.3 mg/kg.
- Exhibits anti-inflammatory activity
- Displays anti-lipoperoxidative activity
- Inhibits rat-brain phospholipid peroxidation
- Reduces carrageenin-induced edema
- Purity of 97.0%
- Appearance: solid
- Color: white to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.