Learn More
Grainger Vestil Laminated Rubber Wheel Chock, Ergonomic Handle, Black/Yellow, 1 ea
Catalog No. 502774711
Encompass_Preferred
This laminated rubber wheel chock features an ergonomic handle to simplify positioning and removal from under trailer wheels. Designed to reduce the risk of back, knee, and foot strain, this heavy-duty chock is suitable for multi-purpose trailer applications.
  • Reinforced rubber construction resists tearing, abrasion, and ozone weathering
  • Ergonomic steel handle includes a yellow powder-coated finish for durability
  • Includes an integrated D-ring for the attachment of a security chain (chain not included)
  • Dimensions: 25.72 cm x 51.28 cm x 88.74 cm (10-1/8 in. x 20-3/16 in. x 34-15/16 in.)
  • Assembly required
Catalog No. 50-277-4711 Supplier Grainger Supplier No. 172X21
This laminated rubber wheel chock features an ergonomic handle to simplify positioning and removal from under trailer wheels. Designed to reduce the risk of back, knee, and foot strain, this heavy-duty chock is suitable for multi-purpose trailer applications.
  • Reinforced rubber construction resists tearing, abrasion, and ozone weathering
  • Ergonomic steel handle includes a yellow powder-coated finish for durability
  • Includes an integrated D-ring for the attachment of a security chain (chain not included)
  • Dimensions: 25.72 cm x 51.28 cm x 88.74 cm (10-1/8 in. x 20-3/16 in. x 34-15/16 in.)
  • Assembly required