Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Laminator Pouches

Catalog No. 19124564 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-124-564 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-124-564 Supplier Brady™ Supplier No. 30428LS
Add to Cart
Add to Cart

Pre-Cut Overlaminate Label - Clear: Pre-cut overlaminate is an easy and affordable way to protect your printed and write-on labels without heat, electricity or messy chemicals.

  • Protects printed and write-on labels from harsh environments and smudging
  • Flexible overlaminate conforms to curved surfaces
  • Made of durable polyester with permanent adhesive
  • Laminator not required - hand applied

Specifications

Product Type Laminate
Color Clear
For Use With (Equipment) Brady Label Printers
Material Polyester
Height (English) 6.5 in.
Autoclavable Not Autoclavable
Description Laminate
Shape Rectangle
Quantity 100/Pk.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.