Learn More
Molecular Dimensions LAMINEX TM GLASS BASE 1 MICRON
SDP
Catalog No. NC0761053
Encompass
Protein Crystallography. LAMINEX GLASS BASE 100 MICRON (Pack of 10) For Lipidic Cubic Phase, the Laminex system is built up from lower support plates of glass or UV transparent plastic with an adhesive spacer. On to this base, glass, UV transparent plates or films can be applied to create 96 chambers for crystal growth.
Catalog No. NC0761053 Supplier Molecular Dimensions Supplier No. MD1150100
Protein Crystallography. LAMINEX GLASS BASE 100 MICRON (Pack of 10) For Lipidic Cubic Phase, the Laminex system is built up from lower support plates of glass or UV transparent plastic with an adhesive spacer. On to this base, glass, UV transparent plates or films can be applied to create 96 chambers for crystal growth.