Learn More
CPC Scientific H-Tyr-Ile-Gly-Ser-Arg-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109627
Encompass
Peptide derived from domain III of the laminin B1 chain represents minimal sequence necessary for cell attachment and laminin receptor-binding activity. It also inhibits lung tumor colony formation in mice injected with melanoma cells. Moreover, this peptide mediates the adhesion of a variety of epithelical cells to laminin.SEQUENCE: H-Tyr-Ile-Gly-Ser-Arg-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: YIGSRMOLECULAR FORMULA: C26H42N8O8MOLECULAR WEIGHT: 594.7STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [110590-64-2]SYNONYMS: Laminin -1 Chain (929-933) (human, mouse), YIGSR, Laminin Fragment (929-933)RESEARCH AREA: CancerREFERENCES:G.Hunt, Exp. Cell Biol., 57, 165 (1989)Y.Iwamoto et al., J. Cell. Physiol., 134, 287 (1988)Y.Iwamoto et al., Science, 238, 1132 (1987) J.Graf et al., Biochemistry, 26, 6896 (1987)SKU(s): LAMN-002A
Catalog No. 50-210-9627 Supplier CPC Scientific Supplier No. LAMN002A
Add to Cart
Add to Cart
Peptide derived from domain III of the laminin B1 chain represents minimal sequence necessary for cell attachment and laminin receptor-binding activity. It also inhibits lung tumor colony formation in mice injected with melanoma cells. Moreover, this peptide mediates the adhesion of a variety of epithelical cells to laminin.SEQUENCE: H-Tyr-Ile-Gly-Ser-Arg-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: YIGSRMOLECULAR FORMULA: C26H42N8O8MOLECULAR WEIGHT: 594.7STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [110590-64-2]SYNONYMS: Laminin -1 Chain (929-933) (human, mouse), YIGSR, Laminin Fragment (929-933)RESEARCH AREA: CancerREFERENCES:G.Hunt, Exp. Cell Biol., 57, 165 (1989)Y.Iwamoto et al., J. Cell. Physiol., 134, 287 (1988)Y.Iwamoto et al., Science, 238, 1132 (1987) J.Graf et al., Biochemistry, 26, 6896 (1987)SKU(s): LAMN-002A