Learn More
Advanced Biomatrix Recombinant Laminin 221, 0.35 mg
Catalog No. 502255943
Encompass_Preferred
Laminin-221 is found predominantly in the basal lamina of striated muscle tissues. Expression deficiencies are implicated in muscular dystrophy and cardiomyopathies. iMatrix-221 is unique in that no pre-coating steps are necessary. This kit comes with 2 vials for a total of 0.35 mg of protein.
This item is not returnable. View return policy
Catalog No. 50-225-5943 Supplier Advanced Biomatrix Supplier No. 53462EA
Laminin-221 is found predominantly in the basal lamina of striated muscle tissues. Expression deficiencies are implicated in muscular dystrophy and cardiomyopathies. iMatrix-221 is unique in that no pre-coating steps are necessary. This kit comes with 2 vials for a total of 0.35 mg of protein.