Learn More
Advanced Biomatrix Recombinant Laminin 411, 0.35 mg
Supplier:  Advanced Biomatrix 53452EA
Encompass_Preferred
Laminin 411 is predominantly in the vascular endothelial basement membrane. Stem cells cultivated on iMatrix-411 are robustly induced to differentiate into endothelial progenitor cells. iMatrix-411 is unique in that no pre-coating steps are necessary. This kit comes with 2 vials for a total of 0.35 mg of protein.
Catalog No. 50-225-5942