Learn More
ACROBiosystems Human Laminin 411 Protein
Catalog No. 502534517
Encompass_Preferred
Laminin 411 is a key component of the basement membrane and is involved in the regulation of cell differentiation. Laminin 411 fragment consists of the C-terminal end of the alpha 4, beta 1, and gamma 1 chains.
Catalog No. 50-253-4517 Supplier ACROBiosystems Supplier No. LATH5263100UG
Laminin 411 is a key component of the basement membrane and is involved in the regulation of cell differentiation. Laminin 411 fragment consists of the C-terminal end of the alpha 4, beta 1, and gamma 1 chains.