Learn More
Diagnostic Biosystems LAMININ Laminin
Catalog No. 502407310
Encompass_Preferred
Laminins are a family of heterotrimeric extracellular glycoproteins consisting of alpha, beta, and gamma chains. Laminins mediate the attachment, migration and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. Laminin is expressed in heart, lung, kidney, muscle, pancreas, retina, and placenta. Very little or no expression is observed in brain and liver.
Catalog No. 50-240-7310 Supplier Diagnostic Biosystems Supplier No. PDM568
Laminins are a family of heterotrimeric extracellular glycoproteins consisting of alpha, beta, and gamma chains. Laminins mediate the attachment, migration and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. Laminin is expressed in heart, lung, kidney, muscle, pancreas, retina, and placenta. Very little or no expression is observed in brain and liver.