Learn More
Advanced Biomatrix Recombinant Laminin 511, 1.05 mg
Catalog No. 502255941
Encompass_Preferred
The laminin 511 isoform is instrumental to the growth and maintenance of hPSCs through its binding to cell receptors a6B1 integrin. iMatrix-511 is unique in that no pre-coating steps are necessary - saving time and money. This kit comes with 6 vials for a total of 1.05 mg of protein.
This item is not returnable. View return policy
Catalog No. 50-225-5941 Supplier Advanced Biomatrix Supplier No. 53446EA
The laminin 511 isoform is instrumental to the growth and maintenance of hPSCs through its binding to cell receptors a6B1 integrin. iMatrix-511 is unique in that no pre-coating steps are necessary - saving time and money. This kit comes with 6 vials for a total of 1.05 mg of protein.