Learn More
Medchemexpress LLC Laminin 521 protein | >95.0% | 100UG
SDP
Catalog No. 5000176446
Encompass_Preferred
Recombinant human laminin-521 produced in HEK293 cells, supplied for use as a defined extracellular matrix substrate to support feeder-free culture and maintenance of human embryonic and induced pluripotent stem cells. The protein promotes cell adhesion, migration, and differentiation and is provided at high purity with recommended cold-storage conditions.
  • Supports feeder-free culture and maintenance of human pluripotent stem cells.
  • Provides defined extracellular matrix for cell adhesion and differentiation.
  • Produced in mammalian HEK293 cells for appropriate post-translational modifications.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Stable for long-term storage at -80°C when aliquoted to avoid freeze-thaw cycles.
Catalog No. 50-001-76446 Supplier Medchemexpress LLC Supplier No. HYP701311100UG
Recombinant human laminin-521 produced in HEK293 cells, supplied for use as a defined extracellular matrix substrate to support feeder-free culture and maintenance of human embryonic and induced pluripotent stem cells. The protein promotes cell adhesion, migration, and differentiation and is provided at high purity with recommended cold-storage conditions.
  • Supports feeder-free culture and maintenance of human pluripotent stem cells.
  • Provides defined extracellular matrix for cell adhesion and differentiation.
  • Produced in mammalian HEK293 cells for appropriate post-translational modifications.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Stable for long-term storage at -80°C when aliquoted to avoid freeze-thaw cycles.