Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CellAdhere laminin-521 | >98.0% | 50UG
SDP

Catalog No. 5000176213
Encompass_Preferred

Laminin 521 Protein, Human (HEK293) is a recombinant heterotrimeric extracellular matrix glycoprotein used as a defined substrate to support adhesion, self-renewal, and differentiation of human pluripotent stem cells. Supplied as a filtered solution for cell culture applications, it is shipped on dry ice and recommended for long-term storage at -80°C.

  • Recombinant human heterotrimeric glycoprotein (α5β2γ1) expressed in HEK293 cells.
  • Supports feeder-free culture and maintenance of hESCs and hiPSCs.
  • High purity as reported by manufacturer (>95% by SEC-HPLC; batch COA ≥98%).
  • Low endotoxin level (<1 EU/mL) suitable for sensitive cell culture.
  • Supplied as a 0.2 μm filtered solution in PBS or PBS with 10% glycerol.
  • Shipped on dry ice; store long term at -80°C and avoid repeated freeze-thaw cycles.
  • Available in multiple package sizes to suit different workflows.

Catalog No. 50-001-76213 Supplier Medchemexpress LLC Supplier No. HYP70131150UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laminin 521 Protein, Human (HEK293) is a recombinant heterotrimeric extracellular matrix glycoprotein used as a defined substrate to support adhesion, self-renewal, and differentiation of human pluripotent stem cells. Supplied as a filtered solution for cell culture applications, it is shipped on dry ice and recommended for long-term storage at -80°C.

  • Recombinant human heterotrimeric glycoprotein (α5β2γ1) expressed in HEK293 cells.
  • Supports feeder-free culture and maintenance of hESCs and hiPSCs.
  • High purity as reported by manufacturer (>95% by SEC-HPLC; batch COA ≥98%).
  • Low endotoxin level (<1 EU/mL) suitable for sensitive cell culture.
  • Supplied as a 0.2 μm filtered solution in PBS or PBS with 10% glycerol.
  • Shipped on dry ice; store long term at -80°C and avoid repeated freeze-thaw cycles.
  • Available in multiple package sizes to suit different workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.