Learn More
Genscript Corporation Laminin 521 Human
Supplier:  Genscript Corporation Z03817100
Encompass
Laminin 521 composed of alpha 5 beta 2 and gamma 1 chains is a reliable matrix for feeder-free culture of human embryonic (hES) and induced pluripotent (hiPS) stem cells When combined with defined maintenance media Laminin 521 consistently supports self-renewal of human hES and hiPS cells without spontaneous differentiation
Catalog No. 50-000-56684