Learn More
Apexbio Technology LLC Laminin (925-933) 110590-60-8 10mg
SDP
Catalog No. 5000063063
Encompass_Preferred
Laminin (925-933) is a synthetic peptide corresponding to residues 925-933 (Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg) in the 1 chain of laminin an extracellular matrix glycoprotein Laminins major non-collagenous constituents of basement membranes consist of three polypeptide chains and and mediate diverse cellular processes such as adhesion migration differentiation signaling and neurite outgrowth The 1 chain contains specific integrin-binding sites that facilitate interaction with cellular receptors regulating cell attachment and chemotaxis and influencing metastatic behaviors of tumor cells This peptide is frequently utilized in biomedical research to investigate receptor-mediated cell-matrix interactions cell adhesion migration assays and cancer metastasis-related studies
Catalog No. 50-000-63063 Supplier Apexbio Technology LLC Supplier No. A102310
Laminin (925-933) is a synthetic peptide corresponding to residues 925-933 (Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg) in the 1 chain of laminin an extracellular matrix glycoprotein Laminins major non-collagenous constituents of basement membranes consist of three polypeptide chains and and mediate diverse cellular processes such as adhesion migration differentiation signaling and neurite outgrowth The 1 chain contains specific integrin-binding sites that facilitate interaction with cellular receptors regulating cell attachment and chemotaxis and influencing metastatic behaviors of tumor cells This peptide is frequently utilized in biomedical research to investigate receptor-mediated cell-matrix interactions cell adhesion migration assays and cancer metastasis-related studies