Learn More
CPC Scientific H-Cys-Ser-Arg-Ala-Arg-Lys-Gln-Ala-Ala-Ser-Ile-Lys-Val-Ala-Val-Ser-Ala-Asp-Arg-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific LAMN001A
Encompass
Peptide that supports neuron cells outgrowth and stimulates neuron-like process formation.SEQUENCE: H-Cys-Ser-Arg-Ala-Arg-Lys-Gln-Ala-Ala-Ser-Ile-Lys-Val-Ala-Val-Ser-Ala-Asp-Arg-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: CSRARKQAASIKVAVSADRMOLECULAR FORMULA: C82H149N31O26S1MOLECULAR WEIGHT: 2017.32STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [123063-31-0]SYNONYMS: RESEARCH AREA: CancerREFERENCES:T. Kanemoto et al., PNAS, 87, 2279 (1990)SKU(s): LAMN-001A, LAMN-001B
Catalog No. 50-210-9625