Learn More
Sigma Aldrich Fine Chemicals Biosciences Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 114956-81-9, MFCD00081739, 1mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L20201MG
Encompass
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane 1-2mg/mL in Tris-buffered saline, 0.2 m filtered, BioReagent, suitable for cell culture
Catalog No. 50-165-6863