Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Laminin, MP Biomedicals™

Catalog No. MP215002701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215002701 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215002701 Supplier MP Biomedicals™ Supplier No. 0215002701
Add to Cart
Add to Cart

Supports cell adhesion, proliferation and differentiation. Laminin, MP Biomedicals, an extracellular matrix glycoprotein, is the major non-collagenous component of basal lamina and an integral component of structural scaffolding in animal tissue. It is us

Laminin, an extracellular matrix (ECM) multidomain trimeric glycoprotein, was initially isolated from mouse Engelbreth-Holm-Swarm (EHS) tumor (laminin-1). Laminins associate with type IV collagen via entactin and perlecan and bind to cell membranes through integrin receptors, dystrogylcan glycoprotein complex and Lutheran blood group glycoprotein. Laminin has active domains for collagen binding, cell adhesion, heparin binding, and neurite outgrowth fragment.

Laminin (approximately 1 × 106 daltons) is a cell adhesion factor.

  • Useful in culturing cells on glass or plastic
  • Significantly facilitates the culture of epithelial and endothelial cells
  • Supports growth and differentiation of many cell types, including epithelial, endothelial, neural, muscle, and liver cells
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied in 0.05M TRIS; 0.15M NaCl; pH 7.4
Name Laminin
Quantity 1 mg
Source Mouse EHS Sarcoma
Storage Requirements Store at -70°C.
Form Frozen
Purity or Quality Grade 95+% pure by SDS-PAGE
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.