Learn More
Cusabio Technology LLC Laminin subunit alpha-2
Catalog No. 5000051556
Encompass_Preferred
This kit detects Laminin subunit alpha-2 (LAMA2) in human serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 0 78 ng/mL to 50 ng/mL with a sensitivity of 0 195 ng/mL enabling precise measurement of low-abundance samples The assay requires 50-100 L of sample per test and can be completed within 1-5 hours Absorbance is measured at 450 nm to determine LAMA2 concentrations making it suitable for research applications requiring reliable quantification of this extracellular matrix protein The standardized protocol ensures consistent performance across biological matrices while maintaining analytical specificity for the target analyte
Catalog No. 50-000-51556 Supplier Cusabio Technology LLC Supplier No. CSBEL012726HU
This kit detects Laminin subunit alpha-2 (LAMA2) in human serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 0 78 ng/mL to 50 ng/mL with a sensitivity of 0 195 ng/mL enabling precise measurement of low-abundance samples The assay requires 50-100 L of sample per test and can be completed within 1-5 hours Absorbance is measured at 450 nm to determine LAMA2 concentrations making it suitable for research applications requiring reliable quantification of this extracellular matrix protein The standardized protocol ensures consistent performance across biological matrices while maintaining analytical specificity for the target analyte