Learn More
Cusabio Technology LLC laminin subunit alpha 3
Catalog No. 5000048298
Encompass_Preferred
This kit detects LAMA3 in serum plasma and tissue homogenates offering a measurable range of 9 4 ng/mL to 600 ng/mL with a sensitivity of 2 35 ng/mL Utilizing a quantitative sandwich ELISA method it delivers precise results within 1 to 5 hours using 50-100 L of sample per test The assay is optimized for absorbance readings at 450 nm enabling reliable quantification of LAMA3 concentrations in diverse biological matrices Designed for flexibility and accuracy it supports research applications requiring robust detection of laminin subunit alpha-3 levels in both clinical and experimental settings
Catalog No. 50-000-48298 Supplier Cusabio Technology LLC Supplier No. CSBEL012727HU
This kit detects LAMA3 in serum plasma and tissue homogenates offering a measurable range of 9 4 ng/mL to 600 ng/mL with a sensitivity of 2 35 ng/mL Utilizing a quantitative sandwich ELISA method it delivers precise results within 1 to 5 hours using 50-100 L of sample per test The assay is optimized for absorbance readings at 450 nm enabling reliable quantification of LAMA3 concentrations in diverse biological matrices Designed for flexibility and accuracy it supports research applications requiring robust detection of laminin subunit alpha-3 levels in both clinical and experimental settings