Learn More
Cayman Chemical Laminin (925-933), Purity: ≥98%, CAS: 110590-60-8, 10 mg
SDP
Catalog No. 502989534
Encompass_Preferred
A peptide fragment of the laminin β1 chain; binds to the laminin receptor at 1 mg/ml; stimulates the attachment of HT-1080 and CHO cells to culture plates at 100 and 300 µg/ml; induces chemotaxis of B16/F10 murine melanoma cells; enhances neurite outgrowth in isolated chick embryo dorsal root ganglia and PC12 cells when conjugated to an agarose hydrogel backbone
Catalog No. 50-298-9534 Supplier Cayman Chemical Supplier No. 3114110 MG
A peptide fragment of the laminin β1 chain; binds to the laminin receptor at 1 mg/ml; stimulates the attachment of HT-1080 and CHO cells to culture plates at 100 and 300 µg/ml; induces chemotaxis of B16/F10 murine melanoma cells; enhances neurite outgrowth in isolated chick embryo dorsal root ganglia and PC12 cells when conjugated to an agarose hydrogel backbone