Learn More
Selleck Chemical LLC Lamivudine 10mg 134678-17-4 GR109714X
SDP
Catalog No. 501361544
Encompass_Preferred
Lamivudine (GR109714X) is a potent nucleoside analog reverse transcriptase inhibitor, used for treatment of chronic HBV and HIV/AIDS. It works by blocking the HIV reverse transcriptase and hepatitis B virus polymerase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1544 Supplier Selleck Chemical LLC Supplier No. S170610MG
Lamivudine (GR109714X) is a potent nucleoside analog reverse transcriptase inhibitor, used for treatment of chronic HBV and HIV/AIDS. It works by blocking the HIV reverse transcriptase and hepatitis B virus polymerase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.