Learn More
Medchemexpress LLC Lamivudine | 134678-17-4 | 100 MG
SDP
Catalog No. 5000405618
Encompass_Preferred
Lamivudine (BCH-189) is an orally active nucleoside reverse transcriptase inhibitor (NRTI). It can inhibit HIV reverse transcriptase 1/2 and also the reverse transcriptase of hepatitis B virus. Lamivudine salicylate can penetrate the central nervous system (CNS). It appears as a solid and has a molecular weight of 229.26. Its identified uses include laboratory chemicals and the manufacture of substances.
  • Orally active nucleoside reverse transcriptase inhibitor
  • Inhibits HIV reverse transcriptase 1/2
  • Inhibits reverse transcriptase of hepatitis B virus
  • Salicylate form can penetrate the CNS
  • Solid appearance
  • Molecular weight of 229.26
Catalog No. 50-004-05618 Supplier Medchemexpress LLC Supplier No. HYB0250100MG
Lamivudine (BCH-189) is an orally active nucleoside reverse transcriptase inhibitor (NRTI). It can inhibit HIV reverse transcriptase 1/2 and also the reverse transcriptase of hepatitis B virus. Lamivudine salicylate can penetrate the central nervous system (CNS). It appears as a solid and has a molecular weight of 229.26. Its identified uses include laboratory chemicals and the manufacture of substances.
  • Orally active nucleoside reverse transcriptase inhibitor
  • Inhibits HIV reverse transcriptase 1/2
  • Inhibits reverse transcriptase of hepatitis B virus
  • Salicylate form can penetrate the CNS
  • Solid appearance
  • Molecular weight of 229.26