Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lamivudine-13C,d2 | 2714473-70-6 | 232.26 g/mol | C7(13C)H9D2N3O3S | 1 MG
SDP

Catalog No. 5000284310
Encompass_Preferred

Lamivudine-13C,d2 is an isotopically labeled form of lamivudine used as a stable-isotope reference and tracer in analytical and research applications. The compound carries 13C and deuterium labels and is provided in small packaged quantities suitable for analytical standards and method development.

  • Isotopically labeled with 13C and deuterium.
  • Intended for use as a stable-isotope reference or internal standard.
  • Suitable for mass spectrometry and LC-MS/MS method development.
  • Provided in small packaged quantities for analytical use.
  • CAS number: 2714473-70-6.
  • Molecular weight 232.26 g/mol.

Catalog No. 50-002-84310 Supplier Medchemexpress LLC Supplier No. HYW6542471MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lamivudine-13C,d2 is an isotopically labeled form of lamivudine used as a stable-isotope reference and tracer in analytical and research applications. The compound carries 13C and deuterium labels and is provided in small packaged quantities suitable for analytical standards and method development.

  • Isotopically labeled with 13C and deuterium.
  • Intended for use as a stable-isotope reference or internal standard.
  • Suitable for mass spectrometry and LC-MS/MS method development.
  • Provided in small packaged quantities for analytical use.
  • CAS number: 2714473-70-6.
  • Molecular weight 232.26 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.