Learn More
Apexbio Technology LLC Lamivudine 134678-17-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062394
Encompass_Preferred
Lamivudine is a nucleoside analog that acts primarily as a reverse transcriptase inhibitor disrupting viral replication by interfering with RNA-dependent DNA polymerase activity It inhibits human immunodeficiency virus type 1 (HIV-1) reverse transcriptase with an IC50 value of approximately 0 316 M Lamivudine also suppresses replication of simian retrovirus (SRV-1 and SRV-2) while exhibiting negligible activity on foamy viruses and amphotropic murine leukemia virus (MLV-A) Due to its antiviral mechanism and dual inhibitory effect against both HIV-1 and hepatitis B virus (HBV) lamivudine is frequently utilized in research involving retroviral infection models particularly HIV-1 and HBV coinfection studies to evaluate viral dynamics antiviral efficacy and biochemical responses
Catalog No. 50-000-62394 Supplier Apexbio Technology LLC Supplier No. A84585.1
Lamivudine is a nucleoside analog that acts primarily as a reverse transcriptase inhibitor disrupting viral replication by interfering with RNA-dependent DNA polymerase activity It inhibits human immunodeficiency virus type 1 (HIV-1) reverse transcriptase with an IC50 value of approximately 0 316 M Lamivudine also suppresses replication of simian retrovirus (SRV-1 and SRV-2) while exhibiting negligible activity on foamy viruses and amphotropic murine leukemia virus (MLV-A) Due to its antiviral mechanism and dual inhibitory effect against both HIV-1 and hepatitis B virus (HBV) lamivudine is frequently utilized in research involving retroviral infection models particularly HIV-1 and HBV coinfection studies to evaluate viral dynamics antiviral efficacy and biochemical responses