Learn More
Selleck Chemical LLC Lamivudine S1706-1g
SDP
Catalog No. 503110039
Encompass_Preferred
Lamivudine is a potent nucleoside analog reverse transcriptase inhibitor used for treatment of chronic HBV and HIV/AIDS It works by blocking the HIV reverse transcriptase and hepatitis B virus polymerase
Catalog No. 50-311-0039 Supplier Selleck Chemical LLC Supplier No. S17061G
Lamivudine is a potent nucleoside analog reverse transcriptase inhibitor used for treatment of chronic HBV and HIV/AIDS It works by blocking the HIV reverse transcriptase and hepatitis B virus polymerase