Learn More
Selleck Chemical LLC Lamivudine S1706-10mM/1mL
SDP
Catalog No. 503111588
Encompass_Preferred
Lamivudine is a potent nucleoside analog reverse transcriptase inhibitor used for treatment of chronic HBV and HIV/AIDS It works by blocking the HIV reverse transcriptase and hepatitis B virus polymerase
Catalog No. 50-311-1588 Supplier Selleck Chemical LLC Supplier No. S170610MM/1ML
Lamivudine is a potent nucleoside analog reverse transcriptase inhibitor used for treatment of chronic HBV and HIV/AIDS It works by blocking the HIV reverse transcriptase and hepatitis B virus polymerase