Learn More
Cayman Chemical Lamivudine, 134678-17-4, 50 mg
SDP
Supplier:  Cayman Chemical 18514500
Encompass_Preferred
Molecular Formula C8H11N3O3S, Purity ≥98%, Formula Weight 229.3, Synonyms: (-)-BCH 189, Epivir™, Epivir-HBV™, 3TC, 2’,3’-dideoxy-3’-Thiacytidine; A cytidine analog that inhibits the reverse transcriptases of HIV1 (IC50 = 45 nM), HIV2, and hepatitis B.
Catalog No. NC1264068