Learn More
Medchemexpress LLC Lamivudine (Standard) | 134678-17-4 | 99.9% | 25 MG
SDP
Catalog No. 5000450538
Encompass_Preferred
Lamivudine (Standard) is the analytical standard for Lamivudine, an orally active nucleoside reverse transcriptase inhibitor (NRTI). It is designed for research and analytical applications and can inhibit HIV reverse transcriptase 1/2 as well as the reverse transcriptase of the hepatitis B virus. Additionally, Lamivudine salicylate has the ability to penetrate the central nervous system (CNS).
  • Analytical standard for research and analytical applications.
  • Orally active.
  • Inhibits HIV reverse transcriptase 1/2.
  • Inhibits hepatitis B virus reverse transcriptase.
  • Salicylate form can penetrate the CNS.
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-004-50538 Supplier Medchemexpress LLC Supplier No. 25MGHYB0250R
Lamivudine (Standard) is the analytical standard for Lamivudine, an orally active nucleoside reverse transcriptase inhibitor (NRTI). It is designed for research and analytical applications and can inhibit HIV reverse transcriptase 1/2 as well as the reverse transcriptase of the hepatitis B virus. Additionally, Lamivudine salicylate has the ability to penetrate the central nervous system (CNS).
  • Analytical standard for research and analytical applications.
  • Orally active.
  • Inhibits HIV reverse transcriptase 1/2.
  • Inhibits hepatitis B virus reverse transcriptase.
  • Salicylate form can penetrate the CNS.
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.