Learn More
Medchemexpress LLC Lamivudine (Standard) | 134678-17-4 | MFCD00869739 | 99.9% | 100 MG
SDP
Catalog No. 5000405388
Encompass_Preferred
Lamivudine (Standard) is the analytical standard of Lamivudine, intended for research and analytical applications. Lamivudine (BCH-189) is an orally active nucleoside reverse transcriptase inhibitor (NRTI) that can inhibit HIV reverse transcriptase 1/2 and the reverse transcriptase of the hepatitis B virus. Lamivudine salicylate can penetrate the CNS.
  • Analytical standard used for assay.
  • Utilized in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
Catalog No. 50-004-05388 Supplier Medchemexpress LLC Supplier No. HYB0250R100MG
Lamivudine (Standard) is the analytical standard of Lamivudine, intended for research and analytical applications. Lamivudine (BCH-189) is an orally active nucleoside reverse transcriptase inhibitor (NRTI) that can inhibit HIV reverse transcriptase 1/2 and the reverse transcriptase of the hepatitis B virus. Lamivudine salicylate can penetrate the CNS.
  • Analytical standard used for assay.
  • Utilized in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.