Learn More
Medchemexpress LLC Lamivudine (Standard) | 134678-17-4 | MFCD00869739 | 99.9% | 50 MG
SDP
Catalog No. 5000406185
Encompass_Preferred
Lamivudine (Standard) is an analytical standard and an orally active nucleoside reverse transcriptase inhibitor (NRTI). It is intended for research and analytical applications.
  • Inhibits HIV reverse transcriptase 1/2
  • Inhibits reverse transcriptase of hepatitis B virus
  • Can penetrate the CNS
Catalog No. 50-004-06185 Supplier Medchemexpress LLC Supplier No. HYB0250R50MG
Lamivudine (Standard) is an analytical standard and an orally active nucleoside reverse transcriptase inhibitor (NRTI). It is intended for research and analytical applications.
  • Inhibits HIV reverse transcriptase 1/2
  • Inhibits reverse transcriptase of hepatitis B virus
  • Can penetrate the CNS