Learn More
Cayman Chemical LamotrIgIn-d3 1mg
SDP
Catalog No. 502915432
Encompass_Preferred
An internal standard for the quantification of lamotrigine by GC- or LC-MS
Catalog No. 50-291-5432 Supplier Cayman Chemical Supplier No. 371961 MG
An internal standard for the quantification of lamotrigine by GC- or LC-MS