Learn More
Enzo Life Sciences Lamotrigine (25mg). CAS: 84057-84-1
SDP
Supplier:  Enzo Life Sciences ALX430160M025
Encompass_Preferred
Lamotrigine blocks voltage-gated sodium channels. Inhibits the release of glutamate. Neuroprotective. Anticonvulsant. Alternative name: 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, LTG, BW-430C. Purity: ≥98% (Assay). Appearance: White to off-white powder. Solubility: Soluble in DMSO (60°C, 20mg/ml) or dimethyl formamide; slightly soluble in anhydrous ethanol or dilute hydrochloric acid; almost insoluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9247