Learn More
American Radiolabeled Chemicals Inc LAMOTRIGINE 3H 50UC
SDP
Catalog No. 50901246
Encompass_Preferred
Lamotrigine-3H(G) 50uCi in ethanol, 1 mCi/ml, S.A. 5-10 Ci/mmol, M.W. 256.09, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-246 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 183050UC
Add to Cart
Add to Cart
Request a Quote
Lamotrigine-3H(G) 50uCi in ethanol, 1 mCi/ml, S.A. 5-10 Ci/mmol, M.W. 256.09, Shipped in Dry Ice, Exclusive