Learn More
Medchemexpress LLC Lamotrigine | 84057-84-1 | 99.97% | 500 MG
SDP
Catalog No. 5000409336
Encompass_Preferred
Lamotrigine is a potent, orally active anticonvulsant and antiepileptic agent. It functions by selectively blocking voltage-gated Na+ channels, which stabilizes presynaptic neuronal membranes and inhibits glutamate release. This product is used for research into conditions such as epilepsy and focal seizures.
  • Potent and orally active anticonvulsant/antiepileptic agent.
  • Selectively blocks voltage-gated Na+ channels.
  • Stabilizes presynaptic neuronal membranes.
  • Inhibits glutamate release.
  • Used for research in epilepsy and focal seizures.
  • High purity of 99.97%.
Catalog No. 50-004-09336 Supplier Medchemexpress LLC Supplier No. HYB0495500MG
Lamotrigine is a potent, orally active anticonvulsant and antiepileptic agent. It functions by selectively blocking voltage-gated Na+ channels, which stabilizes presynaptic neuronal membranes and inhibits glutamate release. This product is used for research into conditions such as epilepsy and focal seizures.
  • Potent and orally active anticonvulsant/antiepileptic agent.
  • Selectively blocks voltage-gated Na+ channels.
  • Stabilizes presynaptic neuronal membranes.
  • Inhibits glutamate release.
  • Used for research in epilepsy and focal seizures.
  • High purity of 99.97%.