Learn More
Medchemexpress LLC Lamotrigine | 84057-84-1 | MFCD00865333 | 99.99% | 250 MG
SDP
Catalog No. 5000406492
Encompass_Preferred
Lamotrigine is the analytical standard of Lamotrigine, intended for research and analytical applications. It is a potent and orally active anticonvulsant or antiepileptic agent that selectively blocks voltage-gated Na+ channels, stabilizing presynaptic neuronal membranes and inhibiting glutamate release. It can be used for the research of epilepsy, focal seizure, and other conditions.
  • Selectively blocks voltage-gated Na+ channels
  • Stabilizes presynaptic neuronal membranes
  • Inhibits glutamate release
  • Used in research for epilepsy and focal seizure
  • Employed as a reference standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
Catalog No. 50-004-06492 Supplier Medchemexpress LLC Supplier No. 250MGHYB0495R
Lamotrigine is the analytical standard of Lamotrigine, intended for research and analytical applications. It is a potent and orally active anticonvulsant or antiepileptic agent that selectively blocks voltage-gated Na+ channels, stabilizing presynaptic neuronal membranes and inhibiting glutamate release. It can be used for the research of epilepsy, focal seizure, and other conditions.
  • Selectively blocks voltage-gated Na+ channels
  • Stabilizes presynaptic neuronal membranes
  • Inhibits glutamate release
  • Used in research for epilepsy and focal seizure
  • Employed as a reference standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.