Learn More
LaMotte™ SMART 3™ Colorimeter Reagent Systems
SDPSCI_ED
Catalog No. S95011 Shop All LaMotte Company Products
Change view
Click to view available options
Type:
Carbohydrazide Reagent
Copper Cuprizone Reagent
Fluoride Reagent
Hydrogen Peroxide LR Reagent
Lead Reagent
Molybdenum HR Reagent, Thioglycolate
Phosphorus Total HR Reagent
Phosphorus Total LR Reagent
Surfactant Reagent
Tolyltriazole Reagent
Range:
0 to 0.9 ppm
0 to 1.5 ppm
0 to 2 ppm
0 to 3.5 ppm
0 to 5 ppm
0 to 8 ppm
0.0 to 50 ppm
0.0 to 70 ppm
1 to 30 ppm
No. of Tests:
25
50
100
Test Method:
Ascorbic acid or digestion
Bromophenol blue
Cuprizone
DPD
Iron reduction
Molybdovanadate or Digestion
PAR
SPADNS
Thioglycolate
UV Oxidation or dichromate
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Range No. of Tests Test Method
S95011 Lead Reagent 0 to 5 ppm 50 PAR
S95003 Fluoride Reagent 0 to 2 ppm 50 SPADNS
S95403 Surfactant Reagent 0 to 8 ppm 50 Bromophenol blue
S95106 Phosphorus Total HR Reagent 0.0 to 70 ppm 25 Molybdovanadate or Digestion
S95015 Molybdenum HR Reagent, Thioglycolate 0.0 to 50 ppm 50 Thioglycolate
S94992 Copper Cuprizone Reagent 0 to 2 ppm 50 Cuprizone
S94998 Carbohydrazide Reagent 0 to 0.9 ppm 100 Iron reduction
S95105 Phosphorus Total LR Reagent 0 to 3.5 ppm 25 Ascorbic acid or digestion
S95114 Tolyltriazole Reagent 1 to 30 ppm 50 UV Oxidation or dichromate
S95005 Hydrogen Peroxide LR Reagent 0 to 1.5 ppm 100 DPD
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. S95011 Supplier LaMotte™ Supplier No. 403101
Add to Cart
Add to Cart
SMART 3™ Colorimeter Reagent Systems for use in water quality testing and environmental studies.
  • Reagents for use in water testing and environmental studies.
Specifications
For Use With (Equipment) SMART 3 Colorimeter
No. of Tests 50
Range 0 to 5 ppm
Test Method PAR
Type Lead Reagent