Description
- Reagents for use in water testing and environmental studies.
Specifications
Specifications
| For Use With (Equipment) | SMART 3 Colorimeter |
| No. of Tests | 25 |
| Range | 3 to 25 ppm |
| Test Method | Chromotropic acid or digestion |
| Type | Nitrogen Total Reagent |