Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Chloride Individual Test Kits
SDPSCI_ED

Catalog No. S94975 Shop All LaMotte Company Products
Change view
Click to view available options
Range:
0 to 200ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
S94975 0 to 200ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94975 Supplier LaMotte™ Supplier No. 4503DR02
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Employs a chromate indicator and silver nitrate titrant.

  • Direct reading titrator
  • Range: 0-200 ppm/4 ppm Cl– 0-20,000 ppm/400 ppm Cl–
  • No. of Tests: 50 at 200 ppm

Specifications

For Use With (Application) Environmental education, aquarium and fish farming
Includes 15mL chloride reagent, 2 x 30mL chloride reagent, 15mL phenolphthalein indicator, 15mL sulfuric acid, Test Tube, 5 to 10 to 15mL, glass, with cap
No. of Tests 50
Range 0 to 200ppm
Test Method Direct-reading titrator
Type Chloride Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.