Learn More
LaMotte™ Chlorine DPD #3 Instrument Grade Tablets
SDP
Catalog No. 23027089 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
100 Pack
Catalog No. Quantity
23-027089 100 Pack
1 options
This item is not returnable. View return policy
Keep out of reach of children. May irritate eyes and skin. Harmful if swallowed.
Catalog No. 23-027089 Supplier LaMotte™ Supplier No. 6197AJ/DEL
This item is not returnable. View return policy
Keep out of reach of children. May irritate eyes and skin. Harmful if swallowed.
Economical choice for your chlorine testing needs
  • Chemical reagent used to test for total chlorine
  • 100 tablets to a box
DPD = N, N-Diethyl-1, 4-phenylene-diamine sulfate
Use with existing LaMotte™ chlorine comparators or colorimeters, including the LTC3000W Series Laboratory Turbidity, Chlorine and Color Meter.
Order Info
Come in a pack of 100.
Specifications
For Use With (Application) Use existing LaMotte™ chlorine comparators or colorimeters
Quantity 100 Pack
Type Chlorine DPD 3 Instrument Grade Tablet