Learn More
LaMotte™ Total Coliform Test Kit - Recreational Water
SDPSCI_ED
Catalog No. S94972 Shop All LaMotte Company Products
Change view
Click to view available options
No. of Tests:
44
Catalog No. No. of Tests
S94972 44
1 options
Catalog No. S94972 Supplier LaMotte™ Supplier No. 5850
Add to Cart
Add to Cart
An easy-to-use, disposable tube method that indicates the presence of Total Coliform Bacteria.
  • The water sample is placed in a tube containing a reagent tablet and incubated at room temperature for 48 hours
  • The tube is then examined for positive indicators of coliform bacteria such as color change and formation of gas bubbles
  • Each kit contains 44 capped tubes each containing a Coliform Test Tablet, and a set of laminated color chart and instruction flashcards
Specifications
Volume (Metric) 100 mL
For Use With (Application) Water monitoring educations
Includes 44 capped tubes each containing a Coliform Test Tablet, and a set of laminated color chart and instruction flashcards.
No. of Tests 44
Type Total Coliform Test Kit
WARNING: Cancer - www.P65Warnings.ca.gov