Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Insta-Test™ Ammonia Test Strips
SDPSCI_ED

Catalog No. S05047 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
25 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05047 25 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05047 Supplier LaMotte™ Supplier No. 3023G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For detection of ammonia in water

  • Range values: 0, 0.5, 1.0, 3.0, 6.0ppm
  • Includes color chart label
  • 25 strips per vial

Specifications

For Use With (Application) For detection of ammonia in water
Includes Color chart label
Quantity 25 Vials
Range 0 to 6 ppm
Type Ammonia Test Strip
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.