Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Reagent Refill
SDPSCI_ED

Catalog No. S98241 Shop All LaMotte Company Products
Change view
Click to view available options
Type:
Chlorine Reagent 2
Hardness Tablets No.6
QAC Indicator
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S98241 QAC Indicator
S98544 Chlorine Reagent 2
S98535 Hardness Tablets No.6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. S98241 Supplier LaMotte™ Supplier No. 6413E
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Reagent Refill is a replacement reagent.

Available in multiple sizes:

  • 500 mL bottle provides multiple tests.
  • Kit size bottle.

Specifications

Volume (Metric) 15 mL
For Use With (Application) Reagent Refill
Packaging Bottle
Type QAC Indicator
Volume (English) 0.507 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.