Learn More
LaMotte™ SMART™ Water Analysis Laboratory
SDPSCI_ED
Catalog No. S94989 Shop All LaMotte Company Products
Change view
Click to view available options
Model:
SCL-04, without pH and conductivity meters
SCL-05, 120 V/60 Hz
SCL-05, Reagent Refill
Type:
Reagent Refill
Water Analysis Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Type
S94989 SCL-04, without pH and conductivity meters Water Analysis Kit
S94987 SCL-05, 120 V/60 Hz Water Analysis Kit
S94988 SCL-05, Reagent Refill Reagent Refill
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S94989 Supplier LaMotte™ Supplier No. 199102
Add to Cart
Add to Cart
Includes
Includes digital meters to measure pH, conductivity, and TDS
The SMART™ Water Analysis Laboratory features a digital colorimeter that analyzes test sample color reactions and provides direct readouts for 15 factors.
  • Portable.
  • Measures 24 water quality parameters for pollution detection, environmental studies, and industrial water and wastes.
  • Provide results directly in ppm for 6 additional factors.
  • Programmed to provide direct readouts for over 40 factors using optional reagent packages.
Specifications
Volume (Metric) 250 mL
For Use With (Application) Pollution detection, environmental studies and industrial water and wastes
For Use With (Equipment) SMART colorimeter
Includes Digital meters to measure pH, conductivity, and TDS
Type Water Analysis Kit
Model SCL-04, without pH and conductivity meters
WARNING: Cancer - www.P65Warnings.ca.gov