Learn More
LaMotte™ Turbidity Test Kit
SDPSCI_ED
Catalog No. S65414 Shop All LaMotte Company Products
Change view
Click to view available options
Type:
Turbidity Refill Reagent
Turbidity Test Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
S65414 Turbidity Test Kit
S65415 Turbidity Refill Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S65414 Supplier LaMotte™ Supplier No. 751901
Add to Cart
Add to Cart
This item is not returnable. View return policy
This visual comparison test kit is designed for use in shallow waters where Secchi disc depth reading is not feasible.
  • Readings are made by using a standard turbidity reagent to match the turbidity of a water sample
  • Measurements are expressed in Jackson Turbidity Units (JTU)
  • Reads from 0-200 JTU in 5 JTU steps
Specifications
For Use With (Application) Food and beverage, drinking water, environmental education, industrial water boiling and cooling, laundry and sanitation, water and wastewater
No. of Tests 50
Range Reads from 0-200 JTU in 5 JTU steps
Type Turbidity Test Kit
WARNING: Cancer - www.P65Warnings.ca.gov