Learn More
Bulbtronics Inc USH-102DH 100W Mercury Arc Lamp Microscopy
SDP
Catalog No. NC1618114 Shop All Bulbtronics Inc. Products
Encompass_Preferred
Lamp, Halogen, Bulbtronics; 120V, 100W; Base: No. 11/Mini candelabra; Shape: T4; Overall L: 3-1/8 in.; Contains mercury
Catalog No. NC1618114 Supplier Bulbtronics Inc Supplier No. 0000294
Lamp, Halogen, Bulbtronics; 120V, 100W; Base: No. 11/Mini candelabra; Shape: T4; Overall L: 3-1/8 in.; Contains mercury