Learn More
Hach Company Lamp assembly (GE 1630) for 1720D and 1720E low-range turbidimeters
Catalog No. NC2128812 Shop All Hach Company Products
Encompass
Lamp assembly using a GE 1630 bulb for replacement in Hach 1720D and 1720E low-range turbidimeters. The bulb is factory burned-in to stabilize light output and is ready for immediate installation. Hach recommends annual replacement under normal use to maintain accurate turbidity readings.
  • Factory burned-in bulb for stable light output.
  • Ready for immediate installation.
  • Helps maintain instrument accuracy when replaced annually under normal use.
  • Designed for low-range turbidity measurements.
This item is not returnable. View return policy
Catalog No. NC2128812 Supplier Hach Company Supplier No. 1895000
Lamp assembly using a GE 1630 bulb for replacement in Hach 1720D and 1720E low-range turbidimeters. The bulb is factory burned-in to stabilize light output and is ready for immediate installation. Hach recommends annual replacement under normal use to maintain accurate turbidity readings.
  • Factory burned-in bulb for stable light output.
  • Ready for immediate installation.
  • Helps maintain instrument accuracy when replaced annually under normal use.
  • Designed for low-range turbidity measurements.