Learn More
World Precision Instrument Replacement Lamp for Z-LITE
SDP
Catalog No. NC1007436
Encompass
  • 150W
  • 3250K
  • 200 hour average life
  • For use with the NOVA, Z-LITE and MSI-150
Catalog No. NC1007436 Supplier World Precision Instrument Supplier No. EKE
  • 150W
  • 3250K
  • 200 hour average life
  • For use with the NOVA, Z-LITE and MSI-150